Learn More
Sigma Aldrich Fine Chemicals Biosciences Worthington LD Series Liqu1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F91431EA
Encompass
LD Series Liquid Nitrogen Dewars are designed for storage of small quantities of liquid nitrogen. High-strength aluminum shell is both durable and light-weight. Super insulation provides advanced thermal protection. Neck tube joins inner and outer container providing maximum durability and minimum nitrogen consumption. Cap and neck tube core provide positive closure.
Catalog No. 50-290-9080