Learn More
Sigma Aldrich Fine Chemicals Biosciences Worthington LD Series Liqu1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7439941EA
Encompass
The LD Series Liquid Nitrogen Dewars are designed to store and dispense small amounts of liquid nitrogen. Provides high thermal efficiencyRibbed high-strength aluminum body magniformed neck tube design and durable paint make these dewars ruggedEasy to operate design features a lightweight snap-on cap and necktube provides a tight closure and easy access Provides maximum holding times with superior vacuum performance and insulationAccessories sold separately
Catalog No. 50-290-8894