Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Echelon Biosciences Research Labs WORTMANNIN 1 mg
SDP

Supplier:  Echelon Biosciences Research Labs B02221MG

Encompass

Wortmannin is a specific, irreversible inhibitor of phosphatidylinositol 3-kinases (IC50 ~ 2-3 nM). It shows similar potency for all classes of PI3Ks.nn


nn

Solubility

nDMSO: 10 mg/mL or EtOH: 1 mg/mL, water: insolublen

Storage

nstore dry at -20oC. Stock solutions should be stored frozen (-20oC).n

Featured in Publications

n1. Takahashi K, Tanaka T, Suzuki K. Directional control of WAVE2 membrane targeting by EB1 and phosphatidylinositol 3,4,5-triphosphate. Cell Signal. 2010;22(3):510-8.nn
nn

Documents

nTechnical Data Sheet This item is non returnable

Catalog No. 50-720-008


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.