Learn More
Eurofins Discoverx Wortmannin, 1 mg. (92-1095)
Supplier:  Eurofins Discoverx 921095
Encompass
Wortmannin is a naturally occurring steroid metabolite that potently inhibits phosphatidylinositol 3-kinases (PI3Ks, IC50=1-10 nM). Also inhibits ProLabelK1 (IC50=24 nM), ProLabelK3 (IC50=49 nM), MTOR, DNAPK, PI4Ks, MLCK, and MAPK at higher concentrations. Helps enhance apoptotic effects by inhibiting the PI3K/Akt cascade and blocking the anti-apoptotic effects of cytokines. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9633