Learn More
Enzo Life Sciences Wortmannin, (1mg), CAS Number: 19545-26-7
SDP
Catalog No. NC9923759
Encompass_Preferred
PI3 kinase inhibitor. Alternative Name: KY 12420. Formula: C23H24O8. MW: 428.4. CAS Number: 19545-26-7. MI: 14: 10053. Purity: ≥98% (HPLC, TLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (50mg/ml), methanol (5mg/ml) or 100% ethanol (25mg/ml). Use/Stability: Make solutions fresh immediately before use. DO NOT STORE SOLUTIONS. Long Term Storage: -20°C
Catalog No. NC9923759 Supplier Enzo Life Sciences Supplier No. BMLST4150001
PI3 kinase inhibitor. Alternative Name: KY 12420. Formula: C23H24O8. MW: 428.4. CAS Number: 19545-26-7. MI: 14: 10053. Purity: ≥98% (HPLC, TLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (50mg/ml), methanol (5mg/ml) or 100% ethanol (25mg/ml). Use/Stability: Make solutions fresh immediately before use. DO NOT STORE SOLUTIONS. Long Term Storage: -20°C