Learn More
Apexbio Technology LLC Wortmannin 19545-26-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061724
Encompass_Preferred
Wortmannin is a selective inhibitor targeting phosphatidylinositol-3-kinase (PI3K) with reported IC50 of approximately 1 9 nM It inhibits PI3K through an ATP-noncompetitive binding mechanism interacting directly with the catalytic region Additionally wortmannin functions as a noncompetitive inhibitor of myosin light chain kinase (MLCK) with an IC50 around 1 9 M thus decreasing myosin light chain phosphorylation and associated smooth muscle contraction In biomedical research wortmannin is commonly employed to dissect the PI3K signaling pathway evaluate smooth muscle contractility and explore therapeutic potentials related to vascular dilation and inflammation modulation
Catalog No. 50-000-61724 Supplier Apexbio Technology LLC Supplier No. A85445.1
Wortmannin is a selective inhibitor targeting phosphatidylinositol-3-kinase (PI3K) with reported IC50 of approximately 1 9 nM It inhibits PI3K through an ATP-noncompetitive binding mechanism interacting directly with the catalytic region Additionally wortmannin functions as a noncompetitive inhibitor of myosin light chain kinase (MLCK) with an IC50 around 1 9 M thus decreasing myosin light chain phosphorylation and associated smooth muscle contraction In biomedical research wortmannin is commonly employed to dissect the PI3K signaling pathway evaluate smooth muscle contractility and explore therapeutic potentials related to vascular dilation and inflammation modulation