Learn More
Selleck Chemical LLC Wortmannin S2758-20mg
SDP
Catalog No. 503109347
Encompass_Preferred
Wortmannin is the first described PI3K inhibitor with IC50 of 3 nM in a cell-free assay with little selectivity within the PI3K family Wortmannin blocks autophagosome formation and potently inhibits DNA-PK/ATM with IC50 of 16 nM and 150 nM in cell-free assays Wortmannin also inhibits PLK1 activity
Catalog No. 50-310-9347 Supplier Selleck Chemical LLC Supplier No. S275820MG
Wortmannin is the first described PI3K inhibitor with IC50 of 3 nM in a cell-free assay with little selectivity within the PI3K family Wortmannin blocks autophagosome formation and potently inhibits DNA-PK/ATM with IC50 of 16 nM and 150 nM in cell-free assays Wortmannin also inhibits PLK1 activity