Learn More
Selleck Chemical LLC Wortmannin 5mg 19545-26-7 KY 12420
SDP
Catalog No. NC0435907
Encompass_Preferred
Wortmannin (KY 12420) is the first described PI3K inhibitor with IC50 of 3 nM in a cell-free assay, with little selectivity within the PI3K family. Wortmannin blocks autophagosome formation and potently inhibits DNA-PK/ATM with IC50 of 16 nM and 150 nM in cell-free assays. Wortmannin also inhibits PLK1 activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0435907 Supplier Selleck Chemical LLC Supplier No. S27585MG
Wortmannin (KY 12420) is the first described PI3K inhibitor with IC50 of 3 nM in a cell-free assay, with little selectivity within the PI3K family. Wortmannin blocks autophagosome formation and potently inhibits DNA-PK/ATM with IC50 of 16 nM and 150 nM in cell-free assays. Wortmannin also inhibits PLK1 activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.