Learn More
Cayman Chemical Wortmannin-Rapamycin Conjugate, Purity: ≥98%, CAS: 1067892-47-0, 1 mg
SDP
Catalog No. 502955095
Encompass_Preferred
Consists of analogs of 17-hydroxy wortmannin and rapamycin conjugated via a prodrug linker, which are released upon hydrolysis of the prodrug linker in vivo; inhibits the growth of tumors in mice better than rapamycin alone
Catalog No. 50-295-5095 Supplier Cayman Chemical Supplier No. 136711 MG
Consists of analogs of 17-hydroxy wortmannin and rapamycin conjugated via a prodrug linker, which are released upon hydrolysis of the prodrug linker in vivo; inhibits the growth of tumors in mice better than rapamycin alone