Learn More
Cayman Chemical WortmnInRapamycIn Conjugat
SDP
Catalog No. 502955634
Encompass_Preferred
Consists of analogs of 17-hydroxy wortmannin and rapamycin conjugated via a prodrug linker, which are released upon hydrolysis of the prodrug linker in vivo; inhibits the growth of tumors in mice better than rapamycin alone
Catalog No. 50-295-5634 Supplier Cayman Chemical Supplier No. 13671500 UG
Consists of analogs of 17-hydroxy wortmannin and rapamycin conjugated via a prodrug linker, which are released upon hydrolysis of the prodrug linker in vivo; inhibits the growth of tumors in mice better than rapamycin alone