Learn More
BUCHI Woulff Bottle, 125 mL
SDP
Catalog No. 05405054 Shop All BUCHI Products
Change view
Click to view available options
Volume (Metric):
125 mL
Catalog No. Volume (Metric)
05-405-054 125 mL
1 options
Catalog No. 05-405-054 Supplier BUCHI Supplier No. 11059905
Add to Cart
Add to Cart
Woulff bottle with a 125 mL capacity and P+G safety coating, holder included.
  • For trapping particles and droplets
  • Can also be used for pressure equalization
  • Plastic and Glass safety coating
  • Holder included
Specifications
Material Borosilicate Glass (3.3)
Type Woulff Flask
For Use With (Application) Trapping Particles/Droplets, and Pressure Equalization
For Use With (Equipment) Vacuum Pump
Includes Holder
Material (Coating) Plastic and Glass
Volume (Metric) 125 mL