Learn More
Med Vet International Dynarex Securestrip Adhesive Wound Closures, Sterile, 1" X 4", 200/Case
Catalog No. 502832401
Encompass_Preferred
Adhesive backed, non-woven wound closure strips provide support following early suture/staple removal. Flexible non-woven material Skin friendly adhesive to minimize skin shearing or blistering Provides secure closure Allows maximum porosity and vapor transmission for rapid healing
Catalog No. 50-283-2401 Supplier Med Vet International Supplier No. 3526
Adhesive backed, non-woven wound closure strips provide support following early suture/staple removal. Flexible non-woven material Skin friendly adhesive to minimize skin shearing or blistering Provides secure closure Allows maximum porosity and vapor transmission for rapid healing