Learn More
Enzo Life Sciences WP-AMC (5mg)
SDP
Catalog No. 502011353
Encompass_Preferred
Fluorogenic substrate likely cleaved by dipeptidyl peptidase IV (DPIV, DPPIV, CD26), DPP8, and other prolyl peptidases. Can be used in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em = 460 nm. Alternative name: Trp-Pro-AMC. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1353 Supplier Enzo Life Sciences Supplier No. BMLP4500005
Fluorogenic substrate likely cleaved by dipeptidyl peptidase IV (DPIV, DPPIV, CD26), DPP8, and other prolyl peptidases. Can be used in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em = 460 nm. Alternative name: Trp-Pro-AMC. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.