Learn More
ERA WATERS CORP WP COLIFORM MICROBE QC STAND
SDP
Catalog No. NC9434834
Encompass
WasteWatR Coliforms Two lyophilized samples consist of one negative sample and one positive to be quantitated for total, fecal and E. coli coliforms.
Catalog No. NC9434834 Supplier ERA WATERS CORP Supplier No. ERA083
WasteWatR Coliforms Two lyophilized samples consist of one negative sample and one positive to be quantitated for total, fecal and E. coli coliforms.