Learn More
IDEXX REFERENCE LABORATORIES Colilert Test (100-test pack)
Catalog No. NC1801456
Encompass
This supplier will only sell to Pharmaceutical and Manufacturing companies. They will not ship to Universities or Hospitals.
The Colilert test simultaneously detects or quantifies both total coliforms and Escherichia coli, with results in 24 hours. The Colilert Test uses proprietary Defined Substrate Technology (DST) to simultaneously detect total coliforms and E. coli. Two nutrient-indicators, ONPG and MUG, are the major sources of carbon in Colilert and can be metabolized by the coliform enzyme -galactosidase and the E. coli enzyme -glucuronidase, respectively. This item is only able to be ordered if customer is a pharmaceutical or manufacturing entity.
Catalog No. NC1801456 Supplier IDEXX REFERENCE LABORATORIES Supplier No. 982716300
This supplier will only sell to Pharmaceutical and Manufacturing companies. They will not ship to Universities or Hospitals.
The Colilert test simultaneously detects or quantifies both total coliforms and Escherichia coli, with results in 24 hours. The Colilert Test uses proprietary Defined Substrate Technology (DST) to simultaneously detect total coliforms and E. coli. Two nutrient-indicators, ONPG and MUG, are the major sources of carbon in Colilert and can be metabolized by the coliform enzyme -galactosidase and the E. coli enzyme -glucuronidase, respectively. This item is only able to be ordered if customer is a pharmaceutical or manufacturing entity.