Learn More
Medchemexpress LLC WP9QY (TFA) | 98.7% | 1340.40 | 500 UG
SDP
Catalog No. 5000423468
Encompass_Preferred
WP9QY TFA is a biologically active cyclic peptide that acts as a TNF-α antagonist. It is designed to mimic the critical tumor necrosis factor (TNF) recognition loop on TNF receptor I, thereby preventing interactions between TNF and its receptor. This TNF antagonist serves as a useful template for developing small molecular inhibitors to prevent inflammatory bone destruction and systemic bone loss in rheumatoid arthritis. This product is for research use only.
  • Biologically active peptide
  • TNF-α antagonist
  • Mimics TNF recognition loop on TNF receptor I
  • Prevents interactions of TNF with its receptor
  • Useful for developing small molecular inhibitors
  • Prevents inflammatory bone destruction in rheumatoid arthritis
  • Prevents systemic bone loss in rheumatoid arthritis
Catalog No. 50-004-23468 Supplier Medchemexpress LLC Supplier No. HYP2612A500UG
WP9QY TFA is a biologically active cyclic peptide that acts as a TNF-α antagonist. It is designed to mimic the critical tumor necrosis factor (TNF) recognition loop on TNF receptor I, thereby preventing interactions between TNF and its receptor. This TNF antagonist serves as a useful template for developing small molecular inhibitors to prevent inflammatory bone destruction and systemic bone loss in rheumatoid arthritis. This product is for research use only.
  • Biologically active peptide
  • TNF-α antagonist
  • Mimics TNF recognition loop on TNF receptor I
  • Prevents interactions of TNF with its receptor
  • Useful for developing small molecular inhibitors
  • Prevents inflammatory bone destruction in rheumatoid arthritis
  • Prevents systemic bone loss in rheumatoid arthritis