Learn More
Medchemexpress LLC Wp9qy Tfa | 99.4% | 1340.40 | 5 MG
SDP
Catalog No. 5000423473
Encompass_Preferred
WP9QY TFA is a biologically active cyclic peptide designed as a TNF-α antagonist. It mimics the critical tumor necrosis factor (TNF) recognition loop on TNF receptor I, thereby preventing interactions of TNF with its receptor. This makes it a useful template for developing small molecular inhibitors to prevent inflammatory bone destruction and systemic bone loss in rheumatoid arthritis.
  • Acts as a TNF-α antagonist
  • Designed to mimic TNF recognition loop on TNF receptor I
  • Prevents TNF interactions with its receptor
  • Useful for developing inhibitors for inflammatory bone destruction
  • Helps prevent systemic bone loss in rheumatoid arthritis
Catalog No. 50-004-23473 Supplier Medchemexpress LLC Supplier No. HYP2612A5MG
WP9QY TFA is a biologically active cyclic peptide designed as a TNF-α antagonist. It mimics the critical tumor necrosis factor (TNF) recognition loop on TNF receptor I, thereby preventing interactions of TNF with its receptor. This makes it a useful template for developing small molecular inhibitors to prevent inflammatory bone destruction and systemic bone loss in rheumatoid arthritis.
  • Acts as a TNF-α antagonist
  • Designed to mimic TNF recognition loop on TNF receptor I
  • Prevents TNF interactions with its receptor
  • Useful for developing inhibitors for inflammatory bone destruction
  • Helps prevent systemic bone loss in rheumatoid arthritis