Learn More
Azzota Corp WPCM-20 Pen Conductivity Meter
Catalog No. 5000887297
Encompass_Preferred
WPCM-20 Pen Type Waterproof conductivity Meter Conductivity Range 0 00 20 00mS/cm (1% F S)
Catalog No. 50-008-87297 Supplier Azzota Corp Supplier No. WPCM20
WPCM-20 Pen Type Waterproof conductivity Meter Conductivity Range 0 00 20 00mS/cm (1% F S)