Learn More
Azzota Corp WPCM-30 Pen Conductivity TDS M
Catalog No. 5000887261
Encompass_Preferred
WPCM-30 Pen Type Waterproof conductivity TDS Meter Conductivity Range 0 00 20 00mS/cm (1% F S) TDS Range 0 10ppt (Max 20ppt 1% F S)
Catalog No. 50-008-87261 Supplier Azzota Corp Supplier No. WPCM30
WPCM-30 Pen Type Waterproof conductivity TDS Meter Conductivity Range 0 00 20 00mS/cm (1% F S) TDS Range 0 10ppt (Max 20ppt 1% F S)