Learn More
Med Vet International McKesson Hand Sanitizing Wipes, 16-3500, 100/Box
Catalog No. 503043785
Encompass_Preferred
Hand Sanitizing Wipe McKesson 100 Count Ethyl Alcohol Wipe Individual Packet Cloth-like material saturated with ethyl alcohol (70 percent) solution kills up to 99.9 percent of germs Individually wrapped McKesson instant hand sanitizing wipes retain moisture and offer convenient dispensing Aloe, glycerin and vitamin E soften skin, preventing drying from alcohol The solution is safe for use on hands before eating or drinking Latex-free hand sanitizing wipes make a suitable substitute for soap and water
Catalog No. 50-304-3785 Supplier Med Vet International Supplier No. 1055597BX
Hand Sanitizing Wipe McKesson 100 Count Ethyl Alcohol Wipe Individual Packet Cloth-like material saturated with ethyl alcohol (70 percent) solution kills up to 99.9 percent of germs Individually wrapped McKesson instant hand sanitizing wipes retain moisture and offer convenient dispensing Aloe, glycerin and vitamin E soften skin, preventing drying from alcohol The solution is safe for use on hands before eating or drinking Latex-free hand sanitizing wipes make a suitable substitute for soap and water