Learn More
World Precision Instrument WPI MICRO-ePORE Holder, Platinum Wire
SDP
Catalog No. 502521482
Encompass
These electrode holders are designed exclusively for use the with MICRO-ePORE™ system, which includes the silver wire electrode holders. The platinum wire electrode holders are an optional accessory. Platinum wire electrode holders are important when you are working with proteins. The silver chloride wire is perfect for work with RNA. However, the silver chloride wire tends to denature proteins, reducing the efficiency of the microinjection solution.
Catalog No. 50-252-1482 Supplier World Precision Instrument Supplier No. 300683PT
These electrode holders are designed exclusively for use the with MICRO-ePORE™ system, which includes the silver wire electrode holders. The platinum wire electrode holders are an optional accessory. Platinum wire electrode holders are important when you are working with proteins. The silver chloride wire is perfect for work with RNA. However, the silver chloride wire tends to denature proteins, reducing the efficiency of the microinjection solution.