Learn More
World Precision Instrument WPI MICRO-ePORE™ Pinpoint Cell Penetrator
SDP
Catalog No. 502521079
Encompass
Pinpoint Cell Penetrator for targeted cell microinjection and increased embryo viability 
The new WPI MICRO-ePORE™ pinpoint cell penetrator is a simple and versatile system that can be used for efficient cell microinjection of a diverse array of compounds and biomolecules into oocytes and pre-implantation stage mammalian embryos. Patent pending Flutter Electrode Technology assists in small, clean, precise membrane penetration without tearing or damaging the membrane when you are working on the microinjection of transgenic animals or cell manipulation.
Platinum wire electrode holders (sold separately) are available!
NOTE: Microinjection pump and other accessories shown in the system image are sold separately.
Catalog No. 50-252-1079 Supplier World Precision Instrument Supplier No. MICROEPORE
Pinpoint Cell Penetrator for targeted cell microinjection and increased embryo viability 
The new WPI MICRO-ePORE™ pinpoint cell penetrator is a simple and versatile system that can be used for efficient cell microinjection of a diverse array of compounds and biomolecules into oocytes and pre-implantation stage mammalian embryos. Patent pending Flutter Electrode Technology assists in small, clean, precise membrane penetration without tearing or damaging the membrane when you are working on the microinjection of transgenic animals or cell manipulation.
Platinum wire electrode holders (sold separately) are available!
NOTE: Microinjection pump and other accessories shown in the system image are sold separately.