Learn More
World Precision Instrument WPI P40 Micro Drill System, 110-220V
SDP
Catalog No. 502521265
Encompass
WPI P40 microdrill is perfect for drilling, grinding, or finishing bone, teeth, and other materials. Its high torque motor runs extremely quietly, even at 40,000 RPM. The product includes a foot switch. The drill holder for a stereotaxic frame is sold separately.
Catalog No. 50-252-1265 Supplier World Precision Instrument Supplier No. 505635
WPI P40 microdrill is perfect for drilling, grinding, or finishing bone, teeth, and other materials. Its high torque motor runs extremely quietly, even at 40,000 RPM. The product includes a foot switch. The drill holder for a stereotaxic frame is sold separately.