Learn More
World Precision Instrument WPI Precision Trinocular Surgical Microscope
SDP
Catalog No. 502538629
Encompass
WPI’s improved precision SurgioScope (now with five magnification steps) is a portable, high quality surgical microscope offering outstanding image quality and value. Incorporating an agile extension arm and excellent working distance objectives, the SurgioScope provides convenient movement and maneuverability necessary for accurate positioning. These important features, together with a high quality optical system, provide sharp image contrast and enhanced large field of vision. The SurgioScope comes fully equipped with a foot-controlled motorized focusing system, normally only found in more expensive surgical microscopes. A unique dual lamp housing enables safe and rapid changing of the lamp during an operation, without the need to power down.
Catalog No. 50-253-8629 Supplier World Precision Instrument Supplier No. PSMT5N
Add to Cart
Add to Cart
Request a Quote
WPI’s improved precision SurgioScope (now with five magnification steps) is a portable, high quality surgical microscope offering outstanding image quality and value. Incorporating an agile extension arm and excellent working distance objectives, the SurgioScope provides convenient movement and maneuverability necessary for accurate positioning. These important features, together with a high quality optical system, provide sharp image contrast and enhanced large field of vision. The SurgioScope comes fully equipped with a foot-controlled motorized focusing system, normally only found in more expensive surgical microscopes. A unique dual lamp housing enables safe and rapid changing of the lamp during an operation, without the need to power down.