Learn More
World Precision Instrument WPI Adjustable Platform For Mouse Stereotaxic
SDP
Catalog No. 502539076
Encompass
  • The platform is 7x10cm
  • Adjustable height of 3.5-4.25cm
  • The platform is compatible with 505375 and 505376 Temperature control systems
Catalog No. 50-253-9076 Supplier World Precision Instrument Supplier No. 505384
  • The platform is 7x10cm
  • Adjustable height of 3.5-4.25cm
  • The platform is compatible with 505375 and 505376 Temperature control systems