Learn More
World Precision Instrument WPI Guinea Pig Adaptor Kit
SDP
Catalog No. 502539175
Encompass
  • The Guinea Pig Adaptor includes the 45° ear bars required for guinea pigs.
  • The nose clamp reaches further back behind the incisors than on the rat adaptors.
  • Capable of both vertical and A-P travel.
  • It is available for use with the WPI Rat Stereotaxic Instruments series.
Catalog No. 50-253-9175 Supplier World Precision Instrument Supplier No. 505240
  • The Guinea Pig Adaptor includes the 45° ear bars required for guinea pigs.
  • The nose clamp reaches further back behind the incisors than on the rat adaptors.
  • Capable of both vertical and A-P travel.
  • It is available for use with the WPI Rat Stereotaxic Instruments series.