Learn More
World Precision Instrument WPI Standard Mouse & Rat Digital Dual Stereotaxic Instrument With Adaptors
SDP
Catalog No. 502539084
Encompass
Like the WPI Rat Stereotaxic Instruments, the WPI Mouse and Rat Stereotaxic Instruments has larger laser engraved vernier scales (accurate to 100 um), 80 mm of ventral-dorsal, medial-lateral and anterior-posterior travel, absolute lock at 90° ventral-dorsal and brass-bushings for electrical grounding. It is available as a digital or motorized model.
Catalog No. 50-253-9084 Supplier World Precision Instrument Supplier No. 505373
Add to Cart
Add to Cart
Request a Quote
Like the WPI Rat Stereotaxic Instruments, the WPI Mouse and Rat Stereotaxic Instruments has larger laser engraved vernier scales (accurate to 100 um), 80 mm of ventral-dorsal, medial-lateral and anterior-posterior travel, absolute lock at 90° ventral-dorsal and brass-bushings for electrical grounding. It is available as a digital or motorized model.