Learn More
World Precision Instrument WPI Swiss Tweezers, 11cm, Straight, Diamond Coated Tips
SDP
Catalog No. 502539349
Encompass
Swiss Microdissecting Forceps have fine tips for precision work in microsurgery, microdissection, and applications requiring fine manipulation of tissue or small objects under a microscope. The tips on WPI Microdissecting Forceps are fine and straight. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. You have a choice of steel they are manufactured from, all of which are suitable for autoclaving or chemical sterilization. Surgical grade stainless steel is the standard alloy for surgical instruments. The acid-resistant, anti-magnetic alloy works well in an MRI environment.
Catalog No. 50-253-9349 Supplier World Precision Instrument Supplier No. 504515
Swiss Microdissecting Forceps have fine tips for precision work in microsurgery, microdissection, and applications requiring fine manipulation of tissue or small objects under a microscope. The tips on WPI Microdissecting Forceps are fine and straight. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. You have a choice of steel they are manufactured from, all of which are suitable for autoclaving or chemical sterilization. Surgical grade stainless steel is the standard alloy for surgical instruments. The acid-resistant, anti-magnetic alloy works well in an MRI environment.