Learn More
World Precision Instrument WPI Swiss Tweezers, 13cm, Antimagnetic Body, Replaceable CF Tips
SDP
Catalog No. 502539348
Encompass
Swiss Tweezers with replaceable carbon fiber tips are great for applications requiring non-metalic, non-marring material is required to prevent damage to delicate or sensitive surfaces. The carbon fiber plastic tips offer a combination of precision and gentleness. These are great for handling electronics components that are sensitive to static electricity, because they are non-conductive. The tips on WPI Swiss Tweezers are pointed. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. The handle is manufactured from an anti-magnetic stainless steel allow, which is suitable for autoclaving or chemical sterilization. These surgical forceps are quality Swiss-made instruments. Replacement carbon fiber tips are also available and sold separately.
Catalog No. 50-253-9348 Supplier World Precision Instrument Supplier No. 504517
Swiss Tweezers with replaceable carbon fiber tips are great for applications requiring non-metalic, non-marring material is required to prevent damage to delicate or sensitive surfaces. The carbon fiber plastic tips offer a combination of precision and gentleness. These are great for handling electronics components that are sensitive to static electricity, because they are non-conductive. The tips on WPI Swiss Tweezers are pointed. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. The handle is manufactured from an anti-magnetic stainless steel allow, which is suitable for autoclaving or chemical sterilization. These surgical forceps are quality Swiss-made instruments. Replacement carbon fiber tips are also available and sold separately.