Learn More
World Precision Instrument WPI Swiss Microdissection Tweezers #7, 11.5cm, Curved Tips, Acid Resistant, Antimagnetic
SDP
Catalog No. 502539352
Encompass
Swiss Microdissecting Forceps have curved fine tips for precision work in microsurgery, microdissection, and applications requiring fine manipulation of tissue or small objects under a microscope. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. They are manufactured from an acid-resistant, anti-magnetic alloy, and they works well in an MRI environment. These surgical forceps are quality Swiss-made instruments. They may be sterilize using an autoclave or by chemical sterilization.
Catalog No. 50-253-9352 Supplier World Precision Instrument Supplier No. 504505
Swiss Microdissecting Forceps have curved fine tips for precision work in microsurgery, microdissection, and applications requiring fine manipulation of tissue or small objects under a microscope. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. They are manufactured from an acid-resistant, anti-magnetic alloy, and they works well in an MRI environment. These surgical forceps are quality Swiss-made instruments. They may be sterilize using an autoclave or by chemical sterilization.