Learn More
World Precision Instrument WPI Swiss Tweezers Set, 5 Pieces
SDP
Catalog No. NC2225232
Encompass
WPI's Swiss Tweezer Set has five pair of tweezes with fine tips for precision work in microsurgery, microdissection, and applications requiring fine manipulation of tissue or small objects under a microscope. Some have fine, straight, pointed tips, one pair has flat, rounded tips, and one pair has fine curved tips. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. They are manufactured from anti-magnetic, acid-resistant stainless steel, which is suitable for autoclaving or chemical sterilization. The acid-resistant, anti-magnetic alloy works well in an MRI environment. All of these surgical forceps are quality Swiss-made instruments.
This item is not returnable. View return policy
Catalog No. NC2225232 Supplier World Precision Instrument Supplier No. 504518
WPI's Swiss Tweezer Set has five pair of tweezes with fine tips for precision work in microsurgery, microdissection, and applications requiring fine manipulation of tissue or small objects under a microscope. Some have fine, straight, pointed tips, one pair has flat, rounded tips, and one pair has fine curved tips. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. They are manufactured from anti-magnetic, acid-resistant stainless steel, which is suitable for autoclaving or chemical sterilization. The acid-resistant, anti-magnetic alloy works well in an MRI environment. All of these surgical forceps are quality Swiss-made instruments.