Learn More
World Precision Instrument High Intensity Fiber Optic Illumination Source-Z-LITE
SDP
Catalog No. NC1267365
Encompass
The Z-LITE Fiber Optic Il­lu­mi­na­tor provides reli­able, un­in­ter­rupt­ed high-intensity light for micro­scopes. Z-LITE allows a continuous range of subdued or concentrated light­ing controlled by a rotary dim­mer on the front panel. Z-LITE may be used with a ring light and single or bi­fur­cat­ed flex­i­ble fiber bundles, en­abling the light beam to be placed ex­act­ly where it is need­ed. Forced air cool­ing prolongs the lamp life. Lamp color temper­a­ture is 3350°K. An interlock switch automatically cuts off power when front panel is opened to replace bulb.
Catalog No. NC1267365 Supplier World Precision Instrument Supplier No. ZLITE
The Z-LITE Fiber Optic Il­lu­mi­na­tor provides reli­able, un­in­ter­rupt­ed high-intensity light for micro­scopes. Z-LITE allows a continuous range of subdued or concentrated light­ing controlled by a rotary dim­mer on the front panel. Z-LITE may be used with a ring light and single or bi­fur­cat­ed flex­i­ble fiber bundles, en­abling the light beam to be placed ex­act­ly where it is need­ed. Forced air cool­ing prolongs the lamp life. Lamp color temper­a­ture is 3350°K. An interlock switch automatically cuts off power when front panel is opened to replace bulb.