Learn More
Medchemexpress LLC HY-103640 10mg Medchemexpress, WR-1065 dihydrochloride CAS:14653-77-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10364010MG
Encompass_Preferred
Medchemexpress, HY-103640 10mg WR-1065 diHCl CAS:14653-77-1 WR-1065 dihydrochloride can protect normal tissues from the toxic effects of certain cancer drugs and activate p53 through a JNK-dependent signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8149