Learn More
Medchemexpress LLC HY-116387 10mg Medchemexpress, WR99210 CAS:47326-86-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11638710MG
Encompass_Preferred
Medchemexpress, HY-116387 10mg WR99210 CAS:47326-86-3 WR99210 is a effective inhibitor of dihydrofolate reductase (DHFR) with an IC50 of <0.075 nM. WR99210 is effective against the most pyrimethamine-resistant Plasmodium falciparum strains[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9813