Learn More
Medchemexpress LLC WR99210 50mg | 47326-86-3 | 394.7 g/mol | C14H18Cl3N5O2 | 50 MG
SDP
Catalog No. 5000227390
Encompass_Preferred
WR99210 is a potent dihydrofolate reductase (DHFR) inhibitor used in antiparasitic research and as a selection reagent for Plasmodium transfection. It is supplied as a research-grade free base and is used in laboratory studies that require selective inhibition of parasite DHFR.
  • Potent dihydrofolate reductase (DHFR) inhibition with subnanomolar activity.
  • Commonly used as a selection reagent for Plasmodium transfection experiments.
  • Provided as the free base form suitable for research applications.
  • Typical pack size: 50 mg.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-27390 Supplier Medchemexpress LLC Supplier No. HY11638750MG
WR99210 is a potent dihydrofolate reductase (DHFR) inhibitor used in antiparasitic research and as a selection reagent for Plasmodium transfection. It is supplied as a research-grade free base and is used in laboratory studies that require selective inhibition of parasite DHFR.
  • Potent dihydrofolate reductase (DHFR) inhibition with subnanomolar activity.
  • Commonly used as a selection reagent for Plasmodium transfection experiments.
  • Provided as the free base form suitable for research applications.
  • Typical pack size: 50 mg.
  • For research use only; not for human or clinical use.