Learn More
Medchemexpress LLC WR99210 hydrochlorid 5mg | 30711-93-4 | 5 MG
SDP
Catalog No. 5000229223
Encompass_Preferred
WR99210 hydrochloride is a potent dihydrofolate reductase (DHFR) inhibitor used in research on Plasmodium spp. and Toxoplasma gondii, and commonly used for selection of Plasmodium transfectants. The compound is supplied as the hydrochloride salt and is intended for laboratory research use only.
  • Potent DHFR inhibitor (IC50 <0.075 nM).
  • Used for antimalarial research and parasite selection.
  • Supplied as a hydrochloride salt for improved solubility.
  • Suitable for storage at -20°C in sealed containers away from moisture.
  • Available in small research quantities for lab use (e.g., 5 mg).
Catalog No. 50-002-29223 Supplier Medchemexpress LLC Supplier No. HY116387A5MG
WR99210 hydrochloride is a potent dihydrofolate reductase (DHFR) inhibitor used in research on Plasmodium spp. and Toxoplasma gondii, and commonly used for selection of Plasmodium transfectants. The compound is supplied as the hydrochloride salt and is intended for laboratory research use only.
  • Potent DHFR inhibitor (IC50 <0.075 nM).
  • Used for antimalarial research and parasite selection.
  • Supplied as a hydrochloride salt for improved solubility.
  • Suitable for storage at -20°C in sealed containers away from moisture.
  • Available in small research quantities for lab use (e.g., 5 mg).