Learn More
GA International Wrap-Around Cryogenic Thermal-Transfer Labels 1" x 1" + 1.72" (wrap) / 25.4mm x 25.4mm + 43.7mm (wrap) Rectangle White + Transparent From -196°C to +100°C (-321°F to +212°F)
Supplier:  GA International HBTT317NOTC11WH
Encompass_Preferred
Self-laminating cryogenic labels that protect the printed information from alcohol, bleach, DMSO, and detergents to ensure the secure identification of tubes and cryo vials. Designed for long-term preservation under cryogenic laboratory conditions, including liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice, and can withstand temperatures up to +100°C (+212°F). Labels can be written on with cryo markers, and printed with all major brands of thermal-transfer printers.
Catalog No. NC1695996