Learn More
GA International Wrap-Around Cryogenic & Autoclave Resistant Self-Laminating Laser Labels 1.125" x 1" + 1.5" (wrap) / 28.6mm x 25.4mm + 38.1mm (wrap) Rectangle White + Transparent From -196°C to +150°C (-321°F to +302°F)
Catalog No. 5000571442
Encompass_Preferred
Cryogenic wrap-around laser labels for long-term cryo-storage in liquid nitrogen and deep freeze harsh environments. Temperature range from -196°C to +150°C. Resistant to autoclave, boiling water, as well as spraying with ethanol, IPA, acetone, and other chemicals and detergents. Cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and gamma irradiation.FDA Compliance: FDA 21 CFR 175.105 for indirect food contact.
Catalog No. 50-005-71442 Supplier GA International Supplier No. DFSL31WH
Cryogenic wrap-around laser labels for long-term cryo-storage in liquid nitrogen and deep freeze harsh environments. Temperature range from -196°C to +150°C. Resistant to autoclave, boiling water, as well as spraying with ethanol, IPA, acetone, and other chemicals and detergents. Cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and gamma irradiation.FDA Compliance: FDA 21 CFR 175.105 for indirect food contact.