Learn More
GA International Wrap-Around Cryogenic & Autoclave Resistant Self-Laminating Laser Labels 1" x 0.6" + 1.6" (wrap) / 25.4mm x 15.2mm + 40.6mm (wrap) Rectangle Yellow + Transparent From -196°C to +150°C (-321°F to +302°F)
Catalog No. 5000571665
Encompass_Preferred
Cryogenic wrap-around laser labels for long-term cryo-storage in liquid nitrogen and deep freeze harsh environments. Temperature range from -196°C to +150°C. Resistant to autoclave, boiling water, as well as spraying with ethanol, IPA, acetone, and other chemicals and detergents. Cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and gamma irradiation.FDA Compliance: FDA 21 CFR 175.105 for indirect food contact.
Catalog No. 50-005-71665 Supplier GA International Supplier No. DFSL65YE
Cryogenic wrap-around laser labels for long-term cryo-storage in liquid nitrogen and deep freeze harsh environments. Temperature range from -196°C to +150°C. Resistant to autoclave, boiling water, as well as spraying with ethanol, IPA, acetone, and other chemicals and detergents. Cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and gamma irradiation.FDA Compliance: FDA 21 CFR 175.105 for indirect food contact.