Learn More
GA International Wrap-Around Cryogenic & Autoclave-Resistant Thermal-Transfer Labels 1.125" x 1" + 1.595" (wrap) / 28.6mm x 25.4mm + 40.5mm (wrap) Rectangle White + Transparent From -196°C to +150°C (-321°F to +302°F)
Catalog No. 5000572299
Encompass_Preferred
Self-laminating autoclave-resistant labels designed for secure identification of samples in cryogenic storage. The wrap-around transparent part of the label securely laminates the printable part of the label and protects against exposure to chemicals and physical damage. Wrap-around labels can withstand temperatures up to 150°C.
Catalog No. 50-005-72299 Supplier GA International Supplier No. CATT303NOTC31WH
Self-laminating autoclave-resistant labels designed for secure identification of samples in cryogenic storage. The wrap-around transparent part of the label securely laminates the printable part of the label and protects against exposure to chemicals and physical damage. Wrap-around labels can withstand temperatures up to 150°C.