Learn More
GA International Wrap-Around Cryogenic & Autoclave-Resistant Thermal-Transfer Labels 1" x 0.625" + 1" (wrap) / 25.4mm x 15.9mm + 25.4mm (wrap) Rectangle White + Transparent From -196°C to +150°C (-321°F to +302°F)
Catalog No. 5000571264
Encompass_Preferred
Self-laminating autoclave-resistant labels designed for secure identification of samples in cryogenic storage. The wrap-around transparent part of the label securely laminates the printable part of the label and protects against exposure to chemicals and physical damage. Wrap-around labels can withstand temperatures up to 150°C.
Catalog No. 50-005-71264 Supplier GA International Supplier No. CATT311NOTC34WH
Self-laminating autoclave-resistant labels designed for secure identification of samples in cryogenic storage. The wrap-around transparent part of the label securely laminates the printable part of the label and protects against exposure to chemicals and physical damage. Wrap-around labels can withstand temperatures up to 150°C.