Learn More
GA International Wrap-Around Cryogenic & Autoclave-Resistant Thermal-Transfer Labels 1" x 1.18" + 1.69" (wrap) / 25.4mm x 30mm + 42.9mm (wrap) Rectangle Green Apple + Transparent From -196°C to +150°C (-321°F to +302°F)
Catalog No. 5000570908
Encompass_Preferred
Self-laminating autoclave-resistant labels designed for secure identification of samples in cryogenic storage. The wrap-around transparent part of the label securely laminates the printable part of the label and protects against exposure to chemicals and physical damage. Wrap-around labels can withstand temperatures up to 150°C.
Catalog No. 50-005-70908 Supplier GA International Supplier No. CATT309NOTC11GA
Self-laminating autoclave-resistant labels designed for secure identification of samples in cryogenic storage. The wrap-around transparent part of the label securely laminates the printable part of the label and protects against exposure to chemicals and physical damage. Wrap-around labels can withstand temperatures up to 150°C.