Learn More
GA International Wrap-Around Cryogenic Thermal-Transfer Labels 1.06" x 0.59" + 0.59" (wrap) / 27mm x 15mm + 15mm (wrap) Rectangle Lavender + Transparent From -196°C to +100°C (-321°F to +212°F)
Catalog No. 5000572395
Encompass_Preferred
Self-laminating cryogenic labels that protect the printed information from alcohol, bleach, DMSO, and detergents to ensure the secure identification of tubes and cryo vials. Designed for long-term preservation under cryogenic laboratory conditions, including liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice, and can withstand temperatures up to +100°C (+212°F). Labels can be written on with cryo markers, and printed with all major brands of thermal-transfer printers.
Catalog No. 50-005-72395 Supplier GA International Supplier No. HBTT339NPNOTC11LA
Add to Cart
Add to Cart
Self-laminating cryogenic labels that protect the printed information from alcohol, bleach, DMSO, and detergents to ensure the secure identification of tubes and cryo vials. Designed for long-term preservation under cryogenic laboratory conditions, including liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice, and can withstand temperatures up to +100°C (+212°F). Labels can be written on with cryo markers, and printed with all major brands of thermal-transfer printers.