Learn More
Med Vet International Cloth Instrument Wrap, 24in x 24in, Each
Catalog No. 502522140
Encompass_Preferred
Autoclavable Wrappers are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability These two-ply wrappers are stitched with a bias binding that is also color-coded for quick identification
Catalog No. 50-252-2140 Supplier Med Vet International Supplier No. W2424
Autoclavable Wrappers are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability These two-ply wrappers are stitched with a bias binding that is also color-coded for quick identification