Learn More
Med Vet International Cloth Instrument Wrap, 45" X 45", Each
Catalog No. 502831181
Encompass_Preferred
Autoclavable Wrappers are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability
Catalog No. 50-283-1181 Supplier Med Vet International Supplier No. W4545
Autoclavable Wrappers are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability