Learn More
Med Vet International Cloth Instrument Wrap, 54" X 54", Each
Catalog No. 502831231
Encompass_Preferred
Autoclavable wrappers are a woven blend of 55% cotton and 45% polyester. Medium weight, 4.1 oz. per square yard, The T-160 thread count and fabric weight ensures durability, These two-ply wrappers are stitched with a bias binding that is also color-coded for quick identification
Catalog No. 50-283-1231 Supplier Med Vet International Supplier No. W5454
Autoclavable wrappers are a woven blend of 55% cotton and 45% polyester. Medium weight, 4.1 oz. per square yard, The T-160 thread count and fabric weight ensures durability, These two-ply wrappers are stitched with a bias binding that is also color-coded for quick identification