Learn More
Med Vet International Cloth Instrument Wrap, 18 in. x 18 in., 6/Pack
Catalog No. 503043885
Encompass_Preferred
Autoclavable Wrap are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability This two-ply wrap is stitched with a bias binding that is also color-coded for quick identification
Catalog No. 50-304-3885 Supplier Med Vet International Supplier No. W1818X6
Autoclavable Wrap are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability This two-ply wrap is stitched with a bias binding that is also color-coded for quick identification