Learn More
Med Vet International Cloth Instrument Wrap, 24" x 24", 3/pack
Catalog No. 503042936
Encompass_Preferred
Autoclavable Wrappers are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability These two-ply wrappers are stitched with a bias binding that is also color-coded for quick identification
Catalog No. 50-304-2936 Supplier Med Vet International Supplier No. W2424X3
Autoclavable Wrappers are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability These two-ply wrappers are stitched with a bias binding that is also color-coded for quick identification