Learn More
Med Vet International Cloth Instrument Wrap, 36inx36in, 5/pack
Catalog No. 501180954
Encompass_Preferred
Autoclavable Wraps are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability These two-ply wrappers are stitched with a bias binding that are also color-coded for quick identification
Catalog No. 50-118-0954 Supplier Med Vet International Supplier No. W3636X5
Autoclavable Wraps are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability These two-ply wrappers are stitched with a bias binding that are also color-coded for quick identification