Learn More
Med Vet International Gemini Super Heavy Instrument Wraps Or Drapes, Single-Ply, 54-in X 54-in, 12/Box
Catalog No. 502831984
Encompass_Preferred
Made with SMS fabric, Gemini wraps offer high-level protection against fluids and particles. Strong wraps offer high-level protection against fluids and particles. Naturally low linting, it helps maintain the integrity of the sterile packs and the surface is friendly to sterilization tape. Ideal for heavy instrument sets, heavy orthopedic and cardiovascular trays which commonly weigh in excess of 20 pounds.
Catalog No. 50-283-1984 Supplier Med Vet International Supplier No. GEM515412
Made with SMS fabric, Gemini wraps offer high-level protection against fluids and particles. Strong wraps offer high-level protection against fluids and particles. Naturally low linting, it helps maintain the integrity of the sterile packs and the surface is friendly to sterilization tape. Ideal for heavy instrument sets, heavy orthopedic and cardiovascular trays which commonly weigh in excess of 20 pounds.